Learn More
Acetylacetaldehyde dimethyl acetal, tech. 90%
Description
Acetaldehyde dimethyl acetal is used as a flavoring agent. It is also used in the preparation of (R)-4,4-dimethoxy-2-butanol, pyrazoles and pyrimidines. Further, it is used in the preparation of N,N-diethyl-[2-(4-fluorophenyl)-5-methylpyrazolo[1,5-a]-pyrimidin-3-yl]acetamide and acetoacetaldehyde.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acetaldehyde dimethyl acetal is used as a flavoring agent. It is also used in the preparation of (R)-4,4-dimethoxy-2-butanol, pyrazoles and pyrimidines. Further, it is used in the preparation of N,N-diethyl-[2-(4-fluorophenyl)-5-methylpyrazolo[1,5-a]-pyrimidin-3-yl]acetamide and acetoacetaldehyde.
Solubility
Miscible with alcohol. Immiscible with water.
Notes
Incompatible with acids and strong oxidizing agents
Specifications
Specifications
| Melting Point | -83°C |
| Density | 0.996 |
| Boiling Point | 170°C to 172°C |
| Flash Point | 57°C (134°F) |
| Refractive Index | 1.415 |
| Quantity | 250 g |
| UN Number | UN1224 |
| Beilstein | 1702372 |
| Solubility Information | Miscible with alcohol. Immiscible with water. |
| Formula Weight | 132.16 |
| Show More |
Safety and Handling
| Hazard Category | H226 |
| Hazard Statement | GHS H Statement H226 Flammable liquid and vapor. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P374-P380-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S. |
| EINECSNumber | 226-605-1 |
| RTECSNumber | EL7592500 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.