Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adeno-associated Virus 9 Mouse anti-Virus, Unconjugated, Clone: HL2372, MilliporeSigma™
Mouse Monoclonal Antibody
$193.50 - $485.50
Specifications
Antigen | Adeno-associated Virus 9 |
---|---|
Clone | HL2372 |
Applications | Dot Blot, ELISA, Neutralization |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF23092MI
|
MilliporeSigma
MABF230925UL |
25 μL |
Each for $193.50
|
|
|||||
MABF23091MI
|
MilliporeSigma
MABF2309100UL |
100 μL |
Each for $485.50
|
|
|||||
Description
Adeno-associated Virus 9 Monoclonal antibody specifically detects Adeno-associated Virus 9 in Virus samples. It is validated for Dot Blot, ELISA, Neutralizing.Specifications
Adeno-associated Virus 9 | |
Dot Blot, ELISA, Neutralization | |
Unconjugated | |
Mouse | |
Virus | |
Adeno-associated virus 9 capsids. | |
Primary | |
Stable for 1 year at -20°C from date of receipt. |
HL2372 | |
Monoclonal | |
Purified | |
Inflammation and Immunology | |
AAV9;Adeno-associated viruses serotype 9 | |
IgG2a | |
Protein G Purified | |
Clone HL2372 is a mouse monoclonal antibody that detects adenoassociated viruses (AAV9 and AAV8). It binds only to intact capsids and not to linear viral proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title