Learn More
Description
Specifications
Specifications
| Antigen | Adenylate Cyclase 3 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:10000-1:20000, Immunocytochemistry/Immunofluorescence 1:10000-1:20000 |
| Formulation | 50% PBS, 50% Glycerol with 5mM Sodium Azide |
| Gene Accession No. | O60266 |
| Gene Alias | AC3EC 4.6.1.1, AC-III, adenylate cyclase 3, adenylate cyclase type 3, Adenylate cyclase type III, Adenylyl cyclase 3, adenylyl cyclase, type III, ATP pyrophosphate-lyase 3, EC 4.6.1, KIAA0511, olfactive type |
| Gene Symbols | ADCY3 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
