Learn More
Promega ADP-Glo™ Kinase Assay + CDK5/p25 Kinase Enzyme System
Catalog No. prv9541 Shop All Promega Corporation Products
Click to view available options
Quantity:
1 Ea.
Recombinant full-length human CDK5 and p25 were co-expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag on both proteins. CDK5 is a member of the Cyclin-Dependent Kinase family that is most abundant in the mammalian brain.
  • Easily Screen and Profile CDK5/p25 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 Ea.
Content And Storage Components at different temperatures
Kit Contents
  • CDK5/p25 Kinase Enzyme System; (V3231) 1 x 10μg
  • ADP-Glo™ Kinase Assay; (V9101) 1 x 1,000 assays
Product Type Kinase Assay + Kinase Enzyme System
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.