Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ago3 Monoclonal Antibody (4B1-F6), Active Motif™

Mouse Monoclonal Antibody
$156.65 - $638.65
Specifications
Antigen | Ago3 |
---|---|
Clone | 4B1-F6 |
Concentration | 1 mg/mL |
Applications | ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-2853
![]() ![]() |
Active Motif
39788 |
10 μg |
Each for $156.65
|
|
|||||
50-199-2852
![]() ![]() |
Active Motif
39787 |
100 μg |
Each for $638.65
|
|
|||||
Description
AGO3 (protein argonaute-3) is required to RNA-mediated gene silencing (RNAi). It binds to short RNAs such as microRNAs (miRNAs) and represses the translation of mRNAs which are complementary to them. AGO3 is proposed to be involved in stabilization of small RNA derivates (riRNA) derived from processed RNA polymerase III-transcribed Alu repeats containing a DR2 retinoic acid response element (RARE) in stem cells and in the subsequent riRNA-dependent degradation of a subset of RNA polymerase II-transcribed coding mRNAs by recruiting a mRNA decapping complex involving EDC4.Specifications
Ago3 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8 | |
AGO3; argonaute 3; argonaute 3, RISC catalytic component; Argonaute RISC catalytic component 3; argonaute RISC catalytic subunit 3; Argonaute3; AW048688; BOS_3617; C130014L07Rik; eIF2C 3; eIF-2C 3; EIF2C1; EIF2C3; Eukaryotic translation initiation factor 2C 3; eukaryotic translation initiation factor 2C, 1; eukaryotic translation initiation factor 2C, 3; hAgo3; mAgo3; Piwi/argonaute family protein meIF2C3; Protein argonaute-3 | |
AGO3 | |
IgG2a | |
Protein G | |
Antibody |
4B1-F6 | |
ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human | |
Q9H9G7 | |
192669 | |
This Ago3 (Argonaute 3) antibody was raised against a recombinant protein corresponding to amino acids 1-175 of human Ago3. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
AGO3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title