Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIDA Antibody, Novus Biologicals™
SDP

Catalog No. NBP178207 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP178207 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP178207 Supplier Novus Biologicals Supplier No. NBP178207
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AIDA Polyclonal specifically detects AIDA in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen AIDA
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-10 mcg/mg
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. NP_073742.2
Gene Alias axin interactor, dorsalization associated, chromosome 1 open reading frame 80, dorsalization-associated protein, FLJ12806, FLJ32421
Gene Symbols AIDA
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 100 and 150 of human Axin Interactor, Dorsalization Associated using the numbering given in entry NP_073742.2 (GeneID 64853).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Developmental Biology
Primary or Secondary Primary
Gene ID (Entrez) 64853
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.