Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIF Antibody (3C11), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB309210 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309210 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309210 Supplier Novus Biologicals Supplier No. NBP250606MFV450
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AIF Monoclonal antibody specifically detects AIF in Human samples. It is validated for Western Blot

Specifications

Antigen AIF
Applications Western Blot
Classification Monoclonal
Clone 3C11
Conjugate mFluor Violet 450 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias AIFapoptosis-inducing factor 1, mitochondrial, apoptosis-inducing factor, mitochondrion-associated, 1, COXPD6, MGC111425, PDCD8striatal apoptosis-inducing factor, programmed cell death 8 (apoptosis-inducing factor), Programmed cell death protein 8
Host Species Mouse
Immunogen AIF-His fusion protein
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, Inhibitors Activators and Regulators, Mitochondrial Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 9131
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.