Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKT1 Antibody (V7), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP20101A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP20101A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP20101A32 Supplier Novus Biologicals Supplier No. NBP250101AF532
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AKT1 Monoclonal specifically detects AKT1 in Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen AKT1
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone V7
Conjugate Alexa Fluor 532
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50 mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AKT, AKT serine/threonine kinase 1, AKT1m, CWS6, EC 2.7.11, EC 2.7.11.1, PKB, PKB alpha, PKBMGC99656, PRKBA, Protein kinase B, Protein Kinase B Alpha, Proto-oncogene c-Akt, RAC, rac protein kinase alpha, RAC-ALPHA, RAC-alpha serine/threonine-protein kinas
Gene Symbols AKT1
Host Species Mouse
Immunogen This AKT1 antibody was made to an ovalbumin-conjugated synthetic peptide KDPMQRLGGG (C-terminal sequence)
Molecular Weight of Antigen 55.7 kDa
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 207
Target Species Zebrafish
Content And Storage Store at 4°C in the dark.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.