Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Albumin and IgG Depletion SpinTrap

Catalog No. 45002176 Shop All Cytiva Products
Catalog No. 45-002-176 Supplier Cytiva Supplier No. 28948020

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Albumin and IgG Depletion SpinTrap are prepacked columns for the depletion of albumin and IgG from human serum and plasma. They are prepacked with high performance SepharoseTM based media with an affinity for human serum albumin (HSA) and IgG

  • Albumin and IgG Depletion SpinTrap is a prepacked column for the depletion of albumin and IgG from human serum or plasma.
  • Albumin and IgG Depletion SpinTrap handles smaller sample volumes (∼50 μl) using a tabletop centrifuge.
  • Columns are prepacked with high performance Sepharose based media with an affinity for human serum albumin (HSA) and IgG.
  • High depletion capacity, removes > 95% HSA and > 90% IgG, with high reproducibility.
  • Simple and fast procedure.
  • Removal of albumin and IgG allows a higher load of less abundant proteins to be included in the analysis and detection of more proteins.
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 7 mm
Flow Rate <4 mL/min
Ligand Type Recombinant Protein G fragment and recombinant Protein binding HAS
Particle Size 34 μm
pH 3 to 9
Volume (Metric) 370 μL
Endcapped No
Product Type Albumin and IgG Depletion
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.