Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALDH1A3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$416.50 - $682.00
Specifications
Antigen | ALDH1A3 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ALDH1A3 Polyclonal specifically detects ALDH1A3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
ALDH1A3 | |
Polyclonal | |
Rabbit | |
Vision | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
aldehyde dehydrogenase 1 family, member A3, Aldehyde dehydrogenase 6, aldehyde dehydrogenase family 1 member A3, ALDH1A6, ALDH6acetaldehyde dehydrogenase 6, EC 1.2.1, EC 1.2.1.5, RalDH3, RALDH-3, Retinaldehyde dehydrogenase 3 | |
ALDH1A3 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse | |
P47895 | |
220 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:ERGGAMATANGAVENGQPDRKPPALPRPIRNLEVKFTKIFIN | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
56 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title