Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Aldolase A/B/C Monoclonal Antibody (E9)

Mouse Monoclonal Antibody
$526.00 - $1800.00
Specifications
Antigen | Aldolase A/B/C |
---|---|
Clone | E9 |
Concentration | 1 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA547377
|
Invitrogen™
MA547377 |
500 μL |
Each for $1,800.00
|
|
|||||
PIMA547476
![]() |
Invitrogen™
MA547476 |
100 μL |
Each for $526.00
|
|
|||||
Description
Aldolase A/B/C Monoclonal Antibody for Western Blot, ICC/IF, IHC
Aldolases are important glycolytic cytosolic enzymes which catalyse the reversible conversion of fructose-1,6-bisphosphate to glyceraldehyde 3-phosphate. There are three aldolase isozymes coded by three distinct genes in mammals, namely aldolases A, B, and C. Aldolase A is heavily expressed in muscle enzyme and aldolase B is heavily expressed in liver and is regarded as a liver specific enzyme. In the adult, aldolase C is the brain-specific isozyme expressed in astrocytes and a few classes of neurons, notably Purkinje cells. Appropriate antibodies to aldolase C are therefore useful to identify astrocytes in cell culture and sectioned material, and the enzyme may be over expressed in some forms of cancer.Specifications
Aldolase A/B/C | |
1 mg/mL | |
Immunohistochemistry, Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 5mM sodium azide | |
P00884, P04075, P05062, P05063, P05064, P05065, P09117, P09972, Q91Y97 | |
11674, 11676, 226, 229, 230, 230163, 24189, 24190, 24191 | |
Recombinant human full-length aldolase C. | |
Antibody |
E9 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Monoclonal | |
Liquid | |
IgG1 | |
Human, Mouse, Rat | |
ALDOC | |
ALDA; ALDB; ALDC | |
ALDOA, ALDOB, ALDOC | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title