Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aldosterone Antibody (BGN/10/42110) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017459 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017459 0.025 mg
NB10062313 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017459 Supplier Novus Biologicals Supplier No. NB100623130.025MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Aldosterone Monoclonal specifically detects Aldosterone in All Species samples. It is validated for ELISA.

Specifications

Antigen Aldosterone
Applications ELISA
Classification Monoclonal
Clone BGN/10/42110
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000
Formulation PBS with 0.09% Sodium Azide
Host Species Mouse
Immunogen Aldosterone (3-CMO) conjugated to Bovine Serum Albumin.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Test Specificity This product is specific for aldosterone and exhibits minimal reactivity with androstenedione, corticosterone, DHEA and desoxycortisol.
Target Species All species
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.