Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha 1 Mannosidase 1A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | alpha 1 Mannosidase 1A |
---|---|
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
alpha 1 Mannosidase 1A Polyclonal specifically detects alpha 1 Mannosidase 1A in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
alpha 1 Mannosidase 1A | |
Polyclonal | |
Rabbit | |
Golgi Apparatus Markers | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
Alpha-1,2-mannosidase IA, EC 3.2.1, EC 3.2.1.113, HUMM3, HUMM9, Man(9)-alpha-mannosidase, MAN9, Man9-mannosidase, Mannosidase alpha class 1A member 1, mannosidase, alpha, class 1A, member 1, mannosyl-oligosaccharide 1,2-alpha-mannosidase IA, Processing alpha-1,2-mannosidase IA | |
MAN1A1 | |
IgG | |
Affinity Purified |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
P33908 | |
4121 | |
This antibody was developed against a recombinant protein corresponding to amino acids: TLQKLPEEIQRDILLEKKKVAQDQLRDKAPFRGLPPVDFVPPIGVESREPADAAIREK | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title