Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-D-Galacturonic Acid Hydrate 95.0+%, TCI America™
SDP

Catalog No. p-7130096 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 91510-62-2
Molecular Formula C6H9O7
Molecular Weight (g/mol) 0.00
MDL Number MFCD00006618
InChI Key AEMOLEFTQBMNLQ-DTEWXJGMSA-M
Synonym d-+-galacturonic acid monohydrate
PubChem CID 91872458
IUPAC Name (2S,3R,4S,5R,6R)-3,4,5,6-tetrahydroxyoxane-2-carboxylate
SMILES *

Specifications

Quantity 5 g
Formula Weight 194.14
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material alpha-D-Galacturonic Acid Hydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.