Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Internexin Antibody, Novus Biologicals™
SDP

Catalog No. NB018136 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018136 0.025 mL
NB300139 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018136 Supplier Novus Biologicals Supplier No. NB3001390.025ML
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 9 publications

alpha-Internexin Polyclonal specifically detects alpha-Internexin in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen alpha-Internexin
Applications Immunohistochemistry, Immunoprecipitation, Western Blot, Immunofluorescence, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:10000 - 1:20000, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1:500 - 1:1000, Immunoprecipitation
Formulation Whole antisera with No Preservative
Gene Accession No. P23565
Gene Alias 66 kDa neurofilament protein, alpha-Inx, FLJ18662, FLJ57501, internexin neuronal intermediate filament protein, alpha, MGC12702, NEF5, Neurofilament 5, neurofilament 5 (66kD), Neurofilament-66, neurofilament-66, tax-binding protein, NF-66alpha-internexin, TXBP-1
Gene Symbols INA
Host Species Rabbit
Immunogen Purified recombinant rat alpha internexin expressed in and purified from E. coli. [UniProt# P23565]
Molecular Weight of Antigen 66 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Cytoskeleton Markers, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 9118
Test Specificity Human and all other mammalian species tested to date.
Target Species Rabbit, Human, Mouse, Bovine, Equine, Rat, Pig
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.