Learn More
Description
Includes
Altair 5 Multigas Detector with data logging, integral pump, rechargeable battery pack, charger, calibration cap, tubing, CD manual
- Thick rubber housing provides non-slip profile for secure grip, easily surviving 304.8cm (10 ft.) drop.
- MotionAlert™ feature activates when a unit does not detect movement for 30 seconds
- MSA exclusive instrument end of sensor life warning
- 40% less calibration gas used per minute than industry average
- Sensor response and clear times in under 15 seconds on LEL, O2, CO, H2S, and SO2 — 50% faster than industry average
- Greater signal stability and repeatability under changing or extreme
- High-resolution monochrome display screen
- Many e-chem sensor options
- Integral pump or diffusion option
- Rechargeable lithium-ion or alkaline battery options
- 17-language display capability
Economy Kits — Monochrome Display
- Chemical
- Construction
- Gas Detection
- General Industry
- Hazardous Materials
- Indoor Air Quality
- Manufacturing
- Nuclear
- Oil Gas
- Telecommunications
- Water/Wastewater
Certifications
- CE ATEX I I 2G
- Ex ia d IIC T4
- UL Class I, Division 1, ABCDFG, Class II, Class III
- CSA C22.2 No. 157 Class I, Div. 1, Groups ABCD
- ANZ Ex ia s I/IIC T4 (Zone 0)
- IEC Ex ia d e I/IIC T4
Warranty and Services
2 years under normal use conditions for LEL/O2/CO/H2S versions
Specifications
Specifications
| Alarm Types | Audible, visual, vibrating |
| Battery Life | 15 hr. |
| Dimensions (L x W x H) | 6.68 x 3.52 x 6.68 in. (16.96 x 8.94 x 16.96cm) |
| Height (English) | 6.68 in. |
| Height (Metric) | 16.96 cm |
| IP Rating | IP65 |
| Length (English) | 6.68 in. |
| Length (Metric) | 16.96 cm |
| Resolution | LEL: 1%, O2: 0.1%, CO: 1ppm, H2S: 1ppm, NO2: 0.1ppm |
| Temperature Range (English) | -4°F to +122°F |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.