Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ ALTAIR™ 5 Multigas Detector Economy Kit

Catalog No. 18039317 Shop All MSA Products
Change view
Click to view available options
Resolution:
LEL: 1%, O2: 0.1%, CO: 1ppm, H2S: 1ppm, ClO2: 0.01ppm
LEL: 1%, O2: 0.1%, CO: 1ppm, H2S: 1ppm, NH3: 0.5ppm
LEL: 1%, O2: 0.1%, CO: 1ppm, H2S: 1ppm, NO2: 0.1ppm
LEL: 1%, O2: 0.1%, CO: 1ppm, PH3: 0.1ppm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Resolution
18-039-317 LEL: 1%, O2: 0.1%, CO: 1ppm, H2S: 1ppm, ClO2: 0.01ppm
18-039-318 LEL: 1%, O2: 0.1%, CO: 1ppm, PH3: 0.1ppm
18-039-315 LEL: 1%, O2: 0.1%, CO: 1ppm, H2S: 1ppm, NH3: 0.5ppm
18-039-314 LEL: 1%, O2: 0.1%, CO: 1ppm, H2S: 1ppm, NO2: 0.1ppm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-039-317 Supplier MSA™ Supplier No. 10094926

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Includes

Altair 5 Multigas Detector with data logging, integral pump, rechargeable battery pack, charger, calibration cap, tubing, CD manual

Featuring a 60 second span calibration and 10 second bump test, this detector is Galaxy™ System-compatible for easy calibration and record keeping

  • Thick rubber housing provides non-slip profile for secure grip, easily surviving 304.8cm (10 ft.) drop.
  • MotionAlert™ feature activates when a unit does not detect movement for 30 seconds
  • MSA exclusive instrument end of sensor life warning
  • 40% less calibration gas used per minute than industry average
  • Sensor response and clear times in under 15 seconds on LEL, O2, CO, H2S, and SO2 — 50% faster than industry average
  • Greater signal stability and repeatability under changing or extreme

  • High-resolution monochrome display screen
  • Many e-chem sensor options
  • Integral pump or diffusion option
  • Rechargeable lithium-ion or alkaline battery options
  • 17-language display capability

Economy Kits — Monochrome Display

  • Chemical
  • Construction
  • Gas Detection
  • General Industry
  • Hazardous Materials
  • Indoor Air Quality
  • Manufacturing
  • Nuclear
  • Oil Gas
  • Telecommunications
  • Water/Wastewater

Certifications

  • CE ATEX I I 2G
  • Ex ia d IIC T4
  • UL Class I, Division 1, ABCDFG, Class II, Class III
  • CSA C22.2 No. 157 Class I, Div. 1, Groups ABCD
  • ANZ Ex ia s I/IIC T4 (Zone 0)
  • IEC Ex ia d e I/IIC T4

Warranty and Services

2 years under normal use conditions for LEL/O2/CO/H2S versions

Specifications

Alarm Types Audible, visual, vibrating
Battery Life 15 hr.
Dimensions (L x W x H) 6.68 x 3.52 x 6.68 in. (16.96 x 8.94 x 16.96cm)
Height (English) 6.68 in.
Height (Metric) 16.96 cm
IP Rating IP65
Length (English) 6.68 in.
Length (Metric) 16.96 cm
Resolution LEL: 1%, O2: 0.1%, CO: 1ppm, H2S: 1ppm, ClO2: 0.01ppm
Temperature Range (English) -4°F to +122°F
Temperature Range (Metric) -20°C to +50°C
Warranty 2 Years
Weight (English) 18.4 oz.
Weight (Metric) 520g
Width (English) 3.52 in.
Width (Metric) 8.94 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.