Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Amersham™ Hybond™ PVDF Membranes: Rolls
GSA_VA

Catalog No. p-4901753 Shop All Cytiva Products
Click to view available options
Dimensions (LxW):
4 m x 254 mm
4 m x 260 mm
Grade:
Hybond LFP
Hybond P
Hybond SEQ

Catalog No. 45-004-021

Supplier: Cytiva 10600021

Only null left
Add to Cart
Add to Cart

Ideal for Western blotting applications where stripping and reprobing are necessary. Cytiva Life Sciences™ Amersham™ Hybond™ PVDF Membranes: Rolls are hydrophobic PVDF membranes.

Supplied in rolls.

  • Hydrophobic PVDF membranes
  • Small pore size eliminates blow-through and increases protein binding over a wide range of molecular weights

Amersham Hybond 0.2μm

  • Compatible with a range of solvents used for rapid destaining
  • Amersham Hybond SEQ 0.2μm
    • Specifically designed for protein sequencing applications and solid phase amino acid analysis
    • Offers excellent blotting for proteins Mr<20,000
    Amersham Hybond LFP 0.2μm
    • Specifically optimized for fluorescence detection in Western blotting applications
    • Provides low background fluorescence, resulting in high sensitivity

    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

    TRUSTED_SUSTAINABILITY

    Specifications

    For Use With (Application) Compatible with a range of solvents used for rapid destaining
    Binding Capacity 363 to 430 μg IgG/cm2 (Incubation Method), 167 to 199 μg BSA/cm2 (Incubation Method)
    Format Roll
    Material PVDF
    Pore Size 0.2 μm
    Dimensions (LxW) 4 m x 260 mm
    Product Line Amersham Hybond
    Grade Hybond P
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.