Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Sheets
GSA_VA

Use for Western blots, dot-blot assays, and other protein or nucleic acid methods. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Sheets offer excellent sensitivity, resolution, and low background.

$213.52 - $924.42

Specifications

For Use With (Application) Western blots, dot-blot assays, and other protein or nucleic acid methods
Format Sheet
View More Specs

Products 13
Catalog Number Mfr. No. Quantity Length (Metric) Width (Metric) Pore Size Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Length (Metric) Width (Metric) Pore Size Price Quantity & Availability  
45-004-025
View Documents
Cytiva
10600033
5/Pk. 60 cm 30 cm 0.45 μm
Pack of 5 for $745.40
Only null left
Add to Cart
 
45-004-024
View Documents
Cytiva
10600032
5/Pk. 60 cm 30 cm 0.2 μm
Pack of 5 for $806.15
Only null left
Add to Cart
 
45-004-031
View Documents
Cytiva
10600041
25/Pk. 20 cm 20 cm 0.45 μm
Pack of 25 for $720.43
Only null left
Add to Cart
 
45-004-032
View Documents
Cytiva
10600042
10/Pk. 20 cm 20 cm 0.45 μm
Pack of 10 for $492.59
Only null left
Add to Cart
 
45-004-033
GSA_VA
View Documents
Cytiva
10600043
25/Pk. 20 cm 20 cm 0.2 μm
Pack of 25 for $788.80
Only null left
Add to Cart
 
45-004-034
View Documents
Cytiva
10600044
10/Pk. 20 cm 20 cm 0.2 μm
Pack of 10 for $468.71
Only null left
Add to Cart
 
45-004-035
View Documents
Cytiva
10600045
25/Pk. 20 cm 20 cm 0.1 μm
Pack of 25 for $924.42
Only null left
Add to Cart
 
45-004-036
View Documents
Cytiva
10600046
10/Pk. 20 cm 20 cm 0.1 μm
Pack of 10 for $590.25
Only null left
Add to Cart
 
45-004-048
View Documents
Cytiva
10600062
25/Pk. 16 cm 14 cm 0.45 μm
Pack of 25 for $450.27
Only null left
Add to Cart
 
45-004-056
View Documents
Cytiva
10600073
10/Pk. 10 cm 10 cm 0.45 μm
Pack of 10 for $213.52
Only null left
Add to Cart
 
45-004-072
View Documents
Cytiva
10600093
25/Pk. 8 cm 9 cm 0.45 μm
Pack of 25 for $250.63
Only null left
Add to Cart
 
45-004-073
View Documents
Cytiva
10600094
25/Pk. 8 cm 9 cm 0.2 μm
Pack of 25 for $221.33
Only null left
Add to Cart
 
45-500-101
View Documents
Cytiva
10600125
10/Pk. 10.2 cm 13.3 cm 0.2 μm
Pack of 10 for $239.79
Only null left
Add to Cart
 
Description

Description

Use for Western blots, dot-blot assays, and other protein or nucleic acid methods. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Sheets offer excellent sensitivity, resolution, and low background.

Individual sheets.

  • 100% pure nitrocellulose
  • High affinity for proteins
  • Blocking ability
  • Compatible with a range of detection methods, including chemiluminescent, chromogenic, and fluorescent

Specifications

Specifications

Western blots, dot-blot assays, and other protein or nucleic acid methods
Sheet
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.