Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMPK alpha 2 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB241960 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB241960 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB241960 Supplier Novus Biologicals Supplier No. NB100238AF700
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AMPK alpha 2 Polyclonal antibody specifically detects AMPK alpha 2 in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen AMPK alpha 2
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias 5'-AMP-activated protein kinase catalytic subunit alpha-2, AMPK subunit alpha-2, AMPK2, AMPK5'-AMP-activated protein kinase, catalytic alpha-2 chain, AMPK-alpha-2 chain, EC 2.7.11, EC 2.7.11.1, PRKAA, protein kinase, AMP-activated, alpha 2 catalytic subunit
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residues 350 and 400 of human 5'-AMP-activated Protein Kinase, catalytic Alpha-2 chain using the numbering given in entry AAH69823.1 (GeneID 5563).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Hypoxia, MAP Kinase Signaling, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction, Translation Control
Primary or Secondary Primary
Gene ID (Entrez) 5563
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.