Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMPK alpha 2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $487.50

Specifications

Antigen AMPK alpha 2
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Regulatory Status RUO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP15632220
SDP
View Documents
Novus Biologicals
NBP15632220UL
20 μL
Each for $206.00
Only null left
Add to Cart
 
NBP156322
SDP
View Documents
Novus Biologicals
NBP156322
100 μL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

AMPK alpha 2 Polyclonal specifically detects AMPK alpha 2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

AMPK alpha 2
Unconjugated
RUO
P54646
5563
Synthetic peptide directed towards the middle region of human PRKAA2 (NP_006243). Peptide sequence: AYHLIIDNRRIMNQASEFYLASSPPSGSFMDDSAMHIPPGLKPHPERMPP.
Primary
62 kDa
Polyclonal
Rabbit
Autophagy, Cancer, Hypoxia, MAP Kinase Signaling, mTOR Pathway, Protein Kinase, Signal Transduction, Translation Control
5'-AMP-activated protein kinase catalytic subunit alpha-2, AMPK subunit alpha-2, AMPK2, AMPK5'-AMP-activated protein kinase, catalytic alpha-2 chain, AMPK-alpha-2 chain, EC 2.7.11, EC 2.7.11.1, PRKAA, protein kinase, AMP-activated, alpha 2 catalytic subunit
PRKAA2
IgG
This product is specific to Subunit or Isoform: alpha-2.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.