Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Andesite Specimen
Shop All Eisco Products

Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Description
- Abundant phenocrysts in fine-grained matrix offer great demonstration of igneous porphyry
- For use in classroom identification labs and exercises
- Correlates well to Next Generation Science Standards Evidence Statements
- Texture and chemistry variation are useful for students learning about differing cooling rates
- The fine-grained felsic matrix contains larger, identifiable hornblende crystals, indicating two different rates of cooling
- Example of porphyry in igneous rocks
Specifications
Specifications
Product Type | Igneous Rock, Andesite |
Quantity | 10/Pk. |
Specimen Size | 2 to 3 cm |
Weight (Metric) | 0.2 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction