Learn More
Annexin II Mouse, Unlabeled, Clone: 5, BD
Mouse Monoclonal Antibody
$286.00 - $566.00
Specifications
Antigen | Annexin II |
---|---|
Clone | 5 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
Annexin II (p36), originally discovered as a substrate for the src oncogene, is one of the most studied members of the annexin family of calcium-dependent phospholipid-binding proteins. It exists in cells as either a monomer or a heterotetramer complexed with p11, an S100-related protein. In vitro, it has been found that unphosphorylated annexin II is capable of causing aggregation of chromaffin granules, and, upon phosphorylation, induces fusion of the granules membranes. A similiar phenomenon may occur in the final steps of exocytosis after cytoplasmic annexin translocates to the subplasmalemmal region and is phosphorylated by protein kinase C. However, other studies refute annexin II's involvement in exocytosis, and a conclusive role for annexin II in membrane trafficking remains debated.
Host Species: Mouse
Clone: 5
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human Annexin II aa. 123-328
Formula Weight [Chemical]: 36kDa
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
Annexin II | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified | |
Lipocortin II, Calpactin I |
5 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Chicken, Human, Mouse, Rat | |
Human Annexin II aa. 123-328 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.