Learn More
APP-BP1 Mouse, Unlabeled, Clone: 20, BD
Mouse Monoclonal Antibody
Specifications
| Antigen | APP-BP1 |
|---|---|
| Clone | 20 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Amyloid precursor protein (APP) gene encodes multiple APPs ranging from 695 to 770 amino acids. These proteins are processed into β-Amyloid peptides (39-43 amino acids), which form abnormal plaques in the cerebral cortex and blood vessel walls during Alzheimer's disease. The transmembrane form of APP is a putative cell surface receptor that possesses neurite-promoting activity, co-localizes with the plaques found in Alzheimer's disease, and is involved in synaptic vesicle recycling. APP binding protein 1 (APP-BP1) interacts with the C-terminus of APP, and is a relative of the ubiquitin-activating enzymes (E1). The structure of APP-BP1 includes a human Uba3 binding site (UBS) at amino acids 443 to 479. This site may be important for APP-BP1 regulation of the cell cycle through interactions with ubiquitinylation-related pathways. Transfection of APP-BP1 in cells with a ts41 mutation suppresses the abnormal S-phases observed in these cells in a hUba3- and hUbc12-dependent manner. In addition, APP-BP1 has been implicated in ubiquitinylation-dependent apoptosis in neurons. Thus, APP-BP1 may be a multi-functional APP-binding protein that regulates cell cycle dynamics.
Host Species: Mouse
Clone: 20
Isotype: IgG2a
Species Reactivity [for Features Main]: Mouse
Immunogen: Human APP-BP1 aa. 348-464
Immunofluorescence, Western Blotting
Specifications
| APP-BP1 | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Human APP-BP1 aa. 348-464 | |
| Primary |
| 20 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Canine, Human, Mouse, Rat | |
| IgG2a | |
| Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.