Learn More
Bcl-2 Mouse, Unlabeled, Clone: 7, BD
Mouse Monoclonal Antibody
$336.00 - $666.00
Specifications
| Antigen | Bcl-2 |
|---|---|
| Clone | 7 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Human hematopoietic malignancies are often characterized by gene rearrangements which deregulate gene expression. The Bcl-2 gene (B-cell leukemia/lymphoma 2) was discovered by walking the chromosomal breakpoints t(14,18). This gene consists of two exons and several transcripts that encode two different protein products, Bcl-2α and Bcl-2β. Both are approximately 26-30kDa and differ at their C-terminal region. Bcl-2 suppresses programmed cell death (apoptosis) and this property appears to be modulated by homodimerization or association with Bax and Bad, two cell death promoters.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
| Bcl-2 | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified |
| 7 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Canine, Chicken, Human, Mouse, Rat | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.