Learn More
Casein Kinase IIα/α Mouse, Unlabeled, Clone: 31, BD
Mouse Monoclonal Antibody
Specifications
| Antigen | Casein Kinase IIα/α |
|---|---|
| Clone | 31 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Casein kinase II (CKII, CK2) is a pleiotropic, ubiquitous, and constitutively active Ser/Thr kinase that utilizes both ATP and GTP as phosphoryl donors to catalyze the phosphorylation of numerous protein substrates. It has been implicated in DNA replication, control of metabolism, and regulation of transcription and translation. CKII is a messenger independent kinase due to the fact that its activity is not affected by factors that regulate other protein kinases (e.g. cAMP, Ca2+, diacylglycerol). It is composed of two catalytic (α and α′) subunits and two regulatory (β) subunits. The β subunits stabilize the holoenzyme and enhance the activity of the α subunits. Although the enzyme exists as a tetramer, the free α/α′ subunits are catalytically active by themselves and are present in cells under some conditions. The α and α′ subunits (85% homologous) are encoded by distinct genes and have been detected in multiple species including human, bovine, and avian. CKII is found primarily in the nucleus in dividing cells, but can be detected in the cytoplasm in quiescent cells. Thus, CKII activity is required for cell cycle progression and other essential signal transduction events.
Host Species: Mouse
Clone: 31
Isotype: IgG1
Species Reactivity [for Features Main]: Rat
Immunogen: Rat Casein Kinase II α aa. 1-123
Immunofluorescence, Western Blotting
Specifications
| Casein Kinase IIα/α | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Rat Casein Kinase II α aa. 1-123 | |
| Primary |
| 31 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| IgG1 | |
| Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.