Learn More
γ-Catenin Mouse, Unlabeled, Clone: 15, BD
Mouse Monoclonal Antibody
$287.50 - $526.00
Specifications
Antigen | γ-Catenin |
---|---|
Clone | 15 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
γ-Catenin (plakoglobin) was identified as a compenent of desmosomes where it associates with desmoglein. γ-Catenin and β-Catenin are closely related proteins that have significant homology with the Drosophila armadillo protein. In addition to complexing with E-Cadherin, γ-Catenin and β-Catenin have been observed in association with the intracellular domain of N-Cadherin. It has been proposed that one molecule of α-Catenin and at least one molecule of β-Catenin and γ-Catenin simultaneously bind to a single cadherin molecule. A 19 amino acid sequence of desmoglein (Dsg1) was found to be critical for binding of γ-Catenin. This region has significant homology to the catenin-binding domain of classical cadherins, thus suggesting a common mechanism for γ-Catenin localization at both adherens junctions and desmosomes.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
γ-Catenin | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG2a | |
Affinity Purified |
15 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Chicken, Human, Mouse, Rat | |
Human γ-Catenin aa. 553-738 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.