Learn More
CD107a Rat anti-Mouse, FITC, Clone: 1D4B, BD
Rat Monoclonal Antibody
$110.00 - $404.00
Specifications
Antigen | CD107a |
---|---|
Clone | 1D4B |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB561069
|
BD Biosciences
561069 |
25 μg |
Each for $110.00
|
|
|||||
BDB553793
|
BD Biosciences
553793 |
0.5 mg |
Each for $404.00
|
|
|||||
Description
The 1D4B antibody recognizes CD107a (Lysosome-Associated Membrane Protein 1, LAMP-1), one of the two major glycoproteins in lysosome membranes which are useful markers to distinguish lysosomes from other organelles. Mouse CD107a consists of a 40kDa core protein which is heavily glycosylated to form heterogeneous mature glycoprotein of 110-140kDa. It is principally expressed in epithelial cells and macrophages in a variety of organs in normal and Beige mutant mice. Several lines of evidence suggest that cell-surface mouse and human LAMP-1 glycoproteins, the expression of which may correlate with malignant transformation, participate in intercellular adhesion and adhesion to extracellular matrix.
Host Species: Rat
Clone: 1D4B
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: Plasma membrane fraction of mouse embryo NIH 3T3 cell line
Intracellular Staining
Specifications
CD107a | |
0.5mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
Plasma membrane fraction of mouse embryo NIH 3T3 cell line | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
1D4B | |
Flow Cytometry | |
FITC | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG2a κ | |
Affinity Purified | |
LAMP-1; LGP-120; LGP-A; P2B |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.