Learn More
anti-CD3zeta/T Cell Receptor zeta Ab-8, Clone: 6B10, Epredia™ LabVision
Mouse Monoclonal Antibody
Specifications
| Antigen | CD3ζ/T Cell Receptor ζ Ab-8 |
|---|---|
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Host Species | Mouse |
| Catalog Number | Mfr. No. | Conjugate | Concentration | Quantity | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Conjugate | Concentration | Quantity | Price | Quantity & Availability | |||
|
MS1289P0
|
Epredia
MS1289P0 |
Unconjugated | 200μg/mL | 100 μL | N/A | N/A | N/A | ||
|
MS1289P1
|
Epredia
MS1289P1 |
Unconjugated | 200μg/mL | 500 μL | N/A | N/A | N/A | ||
|
MS1289P
|
Epredia
MS1289P |
Unconjugated | 200μg/mL | 1 mL | N/A | N/A | N/A | ||
|
MS1289P1ABX
|
Epredia
MS1289P1ABX |
Unconjugated | 1mg/mL | 100 μL | N/A | N/A | N/A | ||
|
MS1289PABX
|
Epredia
MS1289PABX |
Unconjugated | 1mg/mL | 200 μL | N/A | N/A | N/A | ||
Description
CD3 consists of five different polypeptide chains (designated as γ, δ, ε, ζ, and η). The CD3 complex is closely associated at the lymphocyte cell surface with the T cell antigen receptor (TCR). It is involved in signal transduction to the T cell interior following antigen recognition, which culminates in cytokine release, cellular proliferation.. CD3 antigen is a highly specific marker for T cells, and is present in majority of T cell neoplasms.
Host Species: Mouse
Clone: 6B10
Isotype: IgG1
Cross Reactivity: Human, Mouse, and Rat. Others-not tested.
Epitope: aa 36-54 of T Cell Receptor zeta Chain
Immunogen: Synthetic peptide corresponding to aa 36-54 of T Cell Receptor ζ Chain.
Molecular Weight: ∽20kDa
Positive Control: Jurkat cells.
Cellular Localization: Cell membrane
Ab-8 reacts with phosphorylated and unphosphorylated forms of of T Cell Receptor zeta Chain. It co-immunoprecipitates the zeta chain associated tyrosine kinase ZAP-70
Immunoprecipitation, Western Blotting
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
| CD3ζ/T Cell Receptor ζ Ab-8 | |
| Unconjugated | |
| Mouse | |
| Synthetic peptide corresponding to aa 36-54 of T Cell Receptor ζ Chain. | |
| Primary |
| Monoclonal | |
| Purified | |
| Human, Mouse, Rat | |
| IgG1 | |
| Jurkat cells |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.