Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRISP-2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP152012 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP152012 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP152012 Supplier Novus Biologicals Supplier No. NBP152012
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

CRISP-2 Polyclonal specifically detects CRISP-2 in Human, Porcine, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen CRISP-2
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1 ug/ml, Peptide ELISA Detection limit 1:8000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_003287.1
Gene Alias Cancer/testis antigen 36, CRISP-2GAPDL5, CT36TSP1, cysteine-rich secretory protein 2, glyceraldehyde-3-phosphate dehydrogenase-like 5, testis specific protein 1 (probe H4-1 p3-1), Testis-specific protein TPX-1, TPX1MGC111136
Gene Symbols CRISP2
Host Species Goat
Immunogen Peptide with sequence C-QHSDPEDRKTSTR corresponding to internal region according to NP_003287.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7180
Test Specificity Reported variants represent identical protein: NP_003287.1, NP_001135879.1, NP_001135889.1, NP_001135907.1, NP_001135880.1.
Target Species Human, Pig, Canine
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.