Learn More
Description
Specifications
Specifications
| Antigen | Glut1 |
| Applications | Western Blot, ChIP Assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), In vitro Assay |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, Simple Western 1:50, Chromatin Immunoprecipitation reported in scientific literature, Flow Cytometry 1 ug/ml. Use reported by customer review, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen 1:200. Use reported in scientific literature, In vitro assay reported in scientific literature (Trachsel V et al), Flow (Intracellular) 1 ug/ml, Chromatin Immunoprecipitation (ChIP) |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Accession No. | P11166 |
| Gene Alias | Choreoathetosis/Spasticity, Episodic (Paroxysmal Choreoathetosis), CSE, DYT17, DYT18, DYT9, EIG12, Glucose transporter type 1, erythrocyte/brain, GLUT-1, GLUT1DS, HepG2 glucose transporter, HTLVR, Human T-Cell Leukemia Virus (I and II) Receptor, MGC141895, MGC141896, PED, solute carrier family 2 (facilitated glucose transporter), member 1, Solute Carrier Family 2 Member 1, solute carrier family 2, facilitated glucose transporter member 1 |
| Gene Symbols | SLC2A1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
