Learn More
GS27 Mouse, Unlabeled, Clone: 25, BD
Mouse Monoclonal Antibody
Specifications
Antigen | GS27 |
---|---|
Clone | 25 |
Concentration | 250μg/mL |
Applications | Immunofluorescence, Western Blot |
Classification | Monoclonal |
Description
Eukaryotic protein trafficking involves the packaging of target molecules into membranous vesicles that bud from a donor compartment, travel to a specific destination, fuse, and release their components into an acceptor compartment. Components of both the vesicle and the synaptic plasma membrane interact to form a fusion complex that mediates vesicle docking and fusion. This fusion complex contains NSF (N-ethyl-maleimide-sensitive factor), SNAPs (soluble NSF attachment proteins), and receptor proteins (SNAREs) that include synaptobrevin, synaptotagmin, syntaxin, and SNAP-25 (synaptosome-associated protein of 25kDa). Interactions between vesicle SNAREs and target membrane SNARES mediates the specificity of docking. Along one pathway of protein trafficking, the Golgi apparatus receives proteins from the ER. These proteins move from cis-Golgi to trans-Golgi, through a stack of cisternae, towards the trans-Golgi network. From here, they are delivered to their proper destination in the cell. GS27 (Golgi SNARE of 27kDa) (membrin) is a Golgi-associated SNARE. It functions in medial-to-trans-Golgi protein movement.
Immunofluorescence, Western Blotting
Specifications
GS27 | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified | |
Golgi SNARE of 27 kDa; Membrin |
25 | |
Immunofluorescence, Western Blot | |
Unconjugated | |
RUO | |
Canine, Human | |
Human GS27 aa. 5-124 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.