Learn More
KSR-1 Mouse, Unlabeled, Clone: 15, BD
Mouse Monoclonal Antibody
Specifications
Antigen | KSR-1 |
---|---|
Clone | 15 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
Proteins of the Ras superfamily play critical roles in the control of normal and neoplastic proliferation. In mammalian cells there are four true Ras proteins (encoded by Ha-ras, N-ras, Ki-rasA, and Ki-rasB). These proteins relay signals from tyrosine kinases at the plasma membrane to the nucleus via a network of Ser/Thr kinases that includes the MAP kinase (Raf-MEK-ERK) pathway. Kinase suppressor of Ras (KSR-1) was discovered in a genetic screen to identify mutations that suppress constitutively active Ras mutants. KSR-1 contains five domains that are conserved areas (CA) in Drosophila, mouse, and human. CA1 is a unique domain, CA2 is a proline-rich domain, CA3 is a cysteine-rich domain, CA4 is a Ser/Thr rich region, and CA5 contains a C-terminal kinase domain similar to that of Raf. In COS7 cells, ceramide induces the autophosphorylation of KSR-1 and KSR-1-induced phosphorylation and activation of Raf. In addition, overexpression of KSR-1 in 293T cells leads to recruitment of MEK to a 700kDa protein complex and translocates MEK from the soluble to the membrane-associated fraction. Thus, KSR-1 may have important kinase and scaffolding activities during Ras-related signaling.
Host Species: Mouse
Clone: 15
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: Mouse KSR-1 aa. 90-203
Formula Weight [Chemical]: 115kDa
Immunofluorescence, Western Blotting
Specifications
KSR-1 | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Mouse KSR-1 aa. 90-203 | |
Primary |
15 | |
Western Blot | |
Unconjugated | |
RUO | |
Mouse, Rat | |
IgG1 | |
Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.