Learn More
Mint1 Mouse, Unlabeled, Clone: 23, BD
Mouse Monoclonal Antibody
Specifications
| Antigen | Mint1 |
|---|---|
| Clone | 23 |
| Concentration | 250μg/mL |
| Applications | Immunofluorescence, Western Blot |
| Classification | Monoclonal |
Description
Neuronal communication via neurotransmitter release is mediated by the synaptic vesicle cycle. The initial step in exocytosis is the docking of the vesicle in the active zone of the plasma membrane. This step is followed by fusion of the vesicle and plasma membrane and exocytosis. Munc18-1, a major brain protein, is essential for exocytosis. It binds the vesicle fusion protein syntaxin, along with Doc2a and 2b, two proteins that associate peripherally with the synaptic vesicle. Munc18-1 is a family member of membrane trafficking proteins. Its function is thought to be mediated by two Munc18-1-interacting proteins termed Mint 1 and Mint 2, which are 50% homologous. They are expressed exclusively in brain and bind Munc18-1 (MID) with high affinity. They contain an N-terminal Munc18-1 interacting domain and C-terminal PTB (pTyr/PIP interaction) and PDZ (membrane protein interaction) domains, suggesting the ability of Mint proteins to link vesicle exocytosis to Tyr phosphorylation and/or localization at synaptic intercellular junctions. Thus, the Mint proteins, along with Munc18-1 and syntaxin, form a multimeric complex that mediates appropriate docking/fusion of synaptic vesicles.
Immunofluorescence, Western Blotting
Specifications
| Mint1 | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Rat Mint 1 aa. 268-377 | |
| Primary |
| 23 | |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| RUO | |
| Mouse, Rabbit, Rat | |
| IgG1 | |
| Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.