Learn More
PARP Mouse anti-Human, Unlabeled, Clone: 7D3-6, BD
Mouse Monoclonal Antibody
$286.00 - $446.00
Specifications
Antigen | PARP |
---|---|
Clone | 7D3-6 |
Concentration | 0.25mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Includes: Purified Mouse Anti-Human PARP, Jurkat Cell Lysate
Description
PARP [Poly(ADP-ribose) polymerase] is a 116kDa nuclear chromatin-associated enzyme that catalyzes the transfer of ADP-ribose units from NAD+ to a variety of nuclear proteins including topoisomerases, histones, and PARP itself. The catalytic activity of PARP is increased in nonapoptotic cells following DNA damage, and PARP is thought to play an important role in mediating the normal cellular response to DNA damage. Additionally, PARP is a target of the caspase protease activity associated with apoptosis. During apoptosis, PARP is cleaved from the 116kDa intact form into 85kDa and 25kDa fragments. This process separates the amino-terminal DNA-binding domain of the enzyme from the carboxy-terminal catalytic domain resulting in the loss of normal PARP function. Although the role of PARP in apoptosis remains to be elucidated, PARP cleavage is considered to be a marker of apoptosis.
Host Species: Mouse
Clone: 7D3-6
Isotype: IgG1
Species Reactivity: Human
Immunogen: Purified human PARP
Flow Cytometry, Immunoprecipitation, Western Blotting
Specifications
PARP | |
0.25mg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Purified human PARP | |
Primary | |
Store undiluted at -20°C. |
7D3-6 | |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
IgG1 | |
Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.