Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PGE2, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP119177 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP119177 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP119177 Supplier Novus Biologicals Supplier No. NBP119177

Rabbit Polyclonal Antibody

PGE2 Polyclonal specifically detects PGE2 in All Species samples. It is validated for ELISA, Radioimmunoassay.

Specifications

Antigen PGE2
Applications Radioimmune Assays (RIA), ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Radioimmunoassay 1:100-1:2000, Enzyme Immunoassay
Formulation Whole antisera with PBS with 0.1% Sodium Azide
Gene Alias PG E2, PGE 2, PGE2, ProstaglandinE2
Host Species Rabbit
Immunogen Prostaglandin E2
Purification Method Unpurified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Prostaglandin E2
Target Species All species
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.