Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCR β chain Hamster anti-Mouse, FITC, Clone: H57-597, BD
Hamster Monoclonal Antibody
$110.00 - $317.50
Specifications
Antigen | TCR β chain |
---|---|
Clone | H57-597 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
The H57-597 antibody reacts with a common epitope of the β; chain of the T-cell Receptor (TCR) complex on α;β; TCR-expressing thymocytes and peripheral T lymphocytes and NK1.1+ thymocytes and NK-T cells of all mouse strains tested. It does not react with γδ TCR-bearing T cells. In the fetal and adult thymus, the TCR β; -chain may form homodimers or pair with the pre-TCR α; -chain on the surface of immature thymocytes before expression of the TCR α; -chain. Plate-bound or soluble H57-597 antibody activates α;β; TCR-bearing T cells, and plate-bound mAb can induce apoptotic death.Specifications
TCR β chain | |
0.5mg/mL | |
Monoclonal | |
Hamster | |
Immunology | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG2 | |
Affinity Purified |
H57-597 | |
Flow Cytometry | |
FITC | |
RUO | |
Mouse | |
TCR affinity-purified from mouse T-cell hybridoma DO-11.10 | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title