Learn More
Eppendorf™ Anticoagulant-Coated Microcentrifuge Tubes
Eppendorf Safe-Lock Tubes are the result of 50 years of continuous improvement and development. The original Safe-Lock tubes protect your sample with excellent centrifugation stability, feature perfect sealing properties, and reliably prevent sample evaporation. Trust in the original Eppendorf Safe-Lock Tubes, because your samples deserve nothing but the best.
Specifications
Autoclavable | Yes |
---|---|
Capacity (Metric) | 1.5mL |
Material | Polypropylene |
Sterility | Sterile |
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
05-407-13C
|
Eppendorf™
022379208 |
Blue | Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. | N/A | |||||
05-407-13D
|
Eppendorf™
022379216 |
Yellow | Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. | N/A | |||||
Description
- Hinged Eppendorf Safe-Lock lid prevents unintentional lid opening during incubation and storage
- Polypropylene provides high resistance to chemicals, mechanical stress and temperature extremes
- Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results
- g-Safe™: Exceptional centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples
- Precise lid sealing for minimal evaporation rates during long-term storage
- Large frosted lid and surface on the side for easy labeling
- Guaranteed functionality from -86 °C to 100 °C
- Autoclavable when open (121 °C, 20 min)
- Available in lot-certified PCR clean, Eppendorf Forensic DNA Grade, and Biopur quality for guaranteed purity
- Available in Eppendorf Protein and DNA LoBind material for maximum recovery of valuable samples
Specifications
Yes | |
1.5mL | |
Polypropylene | |
Sterile |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.