Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APAF-1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056074T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056074T 0.025 mL
NB10056074 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056074T Supplier Novus Biologicals Supplier No. NB10056074SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

APAF-1 Polyclonal specifically detects APAF-1 in Human, Mouse, Rat, Gerbil samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen APAF-1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen
Formulation Neat whole antisera with 0.05% Sodium Azide
Gene Accession No. O14727
Gene Alias APAF-1apoptotic protease activating factor 1, apoptotic peptidase activating factor, apoptotic peptidase activating factor 1, apoptotic protease activating factor, apoptotic protease-activating factor 1, CED4, DKFZp781B1145, KIAA0413
Gene Symbols APAF1
Host Species Rabbit
Immunogen Full-length recombinant human Apaf1 protein was used as immunogen.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Mitochondrial Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 317
Test Specificity Apaf 1
Target Species Human, Mouse, Rat, Gerbil
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.