Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APEX2 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056625 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056625 0.2 mL
NB10056625T 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056625 Supplier Novus Biologicals Supplier No. NB10056625
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

APEX2 Polyclonal specifically detects APEX2 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation (ChIP).

Specifications

Antigen APEX2
Applications Western Blot, ChIP Assay
Classification Polyclonal
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Chromatin Immunoprecipitation reported in scientific literature (PMID 22761421), Chromatin Immunoprecipitation (ChIP)
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q9UBZ4
Gene Alias AP endonuclease 2, APE2, APEX nuclease (apurinic/apyrimidinic endonuclease) 2, APEX nuclease 2, APEXL2, Apurinic-apyrimidinic endonuclease 2, DNA-(apurinic or apyrimidinic site) lyase 2, DNA-apurinic or apyrimidinic site lyase 2, EC 3.1, EC 4.2.99.18, XTH2
Gene Symbols APEX2
Host Species Rabbit
Immunogen This antibody was generated by immunizing rabbits with EELRALDSEGRALLTQHKI (aa122-140) and GQAKTSEAKDEKELRTSF (aa438-455) peptides of human APEX2.
Molecular Weight of Antigen 57 kDa
Purification Method Protein G purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27301
Test Specificity In HeLa, a 54 kDa band is observed.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.