Learn More
Applied Biosystems™ SYBR™ Green Universal Master Mix
Everything needed for SYBR™ Green dye-based PCR amplification and detection in a convenient, single-tube format.
Supplier: Applied Biosystems™ 4309155
Includes: 2X mixture of SYBR Green I Dye, AmpliTaq Gold DNA Polymerase, dNTPs with dUTP, Passive Reference 1 (ROX), and optimized buffer components.
Description
Applied Biosystems™ SYBR™ Green Universal Master Mix combines SYBR™ Green I dye, AmpliTaq Gold™ DNA Polymerase, dNTPs with dUTP, Passive Reference 1, and optimized buffer in the convenience of a single vial.
- SYBR™ Green I dye detects double-stranded DNA, so specific probes are not required
- AmpliTaq Gold™ DNA polymerase minimizes nonspecific product formation to achieve superior performance
- dUTP significantly reduces carryover contamination when used in conjunction with uracil-DNA glycosylase
- Proprietary buffer enhancements ensure performance and reliability
- Premixed components stored at 2° to 8°C significantly reduce assay setup time
Maximum Flexibility and Convenience:
- Applied Biosystems™ SYBR™ Green Universal Master Mix provides maximum flexibility at reduced cost because no target-specific TaqMan™ probes are required
- SYBR™ Green I dye is a double-stranded DNA binding dye that detects any double-stranded DNA generated during PCR
- The hot-start enzyme AmpliTaq Gold™ DNA Polymerase minimizes nonspecific product formation (including primer-dimers), yielding superior performance and sensitivity
- Passive Internal Reference 1 is provided to normalize non-PCR—related fluorescence fluctuations
- This minimizes well-to-well variability that can result from a variety of causes, such as pipetting error or sample evaporation
- SYBR™ Green I dye is ideal for target identification (screening assays) or when a limited number of assays is needed.
- Store at -20°C
Order Info
Shipping Condition: Wet Ice
Specifications
The 1 x 5 mL vials contain a 2X mixture of SYBR Green 1 Dye, AmpliTaq Gold™ DNA Polymerase, dNTPs with dUTP, Passive Reference 1 (ROX), and optimized buffer components. Sufficient reagents provided for 200 reactions based on a 50 μl reaction volume. Store at -20°C. Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). |
|
Tube | |
qPCR | |
Standard | |
SYBR Green | |
SYBR™ | |
1 x 5 mL | |
200 Reactions of 50 μL | |
2X | |
DNA (Genomic), cDNA |
SYBR | |
High | |
AmpliTaq Gold DNA Polymerase | |
7000 System, 7300 System, 7500 System, 7700 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, QuantStudio™, StepOne™, Standard Mode, StepOnePlus™, Standard Mode, ViiA™ 7 System | |
ROX (Pre-mixed) | |
Real Time PCR SYBR Master Mix | |
Wet Ice | |
Gene Expression | |
200 Reactions | |
1 x 5 mL |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.