Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ TaqMan™ Fast Advanced Master Mix for qPCR

Catalog No. 4444965
Encompass_Preferred
Click to view available options
Quantity:
1 x 1 mL
1 x 5 mL
2 x 5 mL
5 x 5 mL
10 x 5 mL
1 x 50 mL

TaqMan Fast Advanced Master Mix delivers accurate quantification and dependability in less time on an array of qPCR instrument platforms.

TaqMan Fast Advanced Master Mix delivers accurate quantification and dependability in less time on an array of qPCR instrument platforms. It provides best in class performance, including the widest quantifiable range, in both single and duplex reactions, even with challenging targets. TaqMan Fast Advanced Master Mix contains AmpliTaq Fast DNA Polymerase, uracil-N-glycosylase (UNG), dNTPs with dUTP, ROX dye (passive reference), and optimized buffer components. It is supplied at a 2X concentration.

Features of the TaqMan Fast Advanced Master Mix include:

  • Best-in-class performance—superior sensitivity, accuracy, dynamic range, and specificity compared to standard mixes in standard mode
  • Engineered for enhanced benchtop stability—stable at room temperature for up to 72 hours in preassembled reactions
  • Optimized for multiplexing—validated for duplexing with exogenous and endogenous internal positive control assays
  • Reduced run times (<40 minutes) on fast and standard instrumentation—optimized for fast mode on fast instruments and fast cycling conditions on standard instruments
  • Seamlessly transitions into your experiments—validated with TaqMan assays for gene expression and microRNAs, and TaqMan array micro-fluidic cards

Best-in-class performance

TaqMan Fast Advanced Master Mix has been designed to provide performance equal to or better than standard master mixes. It has been benchmarked against the leading suppliers' standard and fast master mixes to help ensure that it succeeds in providing best-in-class sensitivity, accuracy, dynamic range, and specificity. TaqMan Fast Advanced Master Mix's dynamic range (up to 7 logs) is the widest in the industry. The impressive sensitivity of the mix is showcased in the figure below which compares the Ct values of TaqMan Fast Advanced Master Mix and TaqMan Universal PCR Master Mix across a panel of gene expression assays.

Benchtop stability for high-throughput handling and convenience

TaqMan Fast Advanced Master Mix has been engineered to retain a high level of performance in preassembled reactions for up to 72 hours. The stability of this mix provides users of high-throughput liquid handling systems the assurance that the results on the first plate will mimic those of the last plate. The figure below shows an assay that was run upon assembly (time 0) and after 72 hours of incubation at 30°C, simulating the most extreme room temperature scenario.

For those with less extreme throughput needs, the enhanced stability of this master mix provides an overall added convenience to your workflow, as you are no longer constrained to immediately running plates upon assembly.

Optimized for multiplexing

We realize that confidence is paramount when it comes to your results. For added confidence in the results in every well, TaqMan Fast Advanced Master Mix has been designed to help deliver accurate results for duplex reactions using an internal positive control (IPC). The figure below shows results for the experimental target β-actin (gene name: ACTB), which was serially diluted and amplified in single-target reactions and duplex reactions.

Validated for microRNA assays

TaqMan Fast Advanced Master Mix has been validated for use with multiple real-time PCR applications, including microRNA assays. The formulation provides high specificity and dynamic range, the two most critical performance attributes that define successful results when working with microRNAs. The data in the figure below demonstrate excellent PCR linearity over a 6-log range of input template.

Reduced run times on standard instrumentation

TaqMan Fast Advanced Master Mix has been optimized for use with both fast and standard instrumentation, enabling researchers who currently own standard instruments to also reap the performance benefits and time savings this mix provides. The figure below showcases the impressive results achieved when using TaqMan Fast Advanced Master Mix under fast thermal cycling conditions on the Applied Biosystems 7300 Real-Time PCR System. The mix has been tested with all Applied Biosystems standard real-time PCR instrumentation (7900HT, 7500, and 7300 systems) to enable success independent of whether or not you own a fast-enabled instrument.

Order Info

Shipping Conditions: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage Contains 10 tubes (5 mL each) 2X TaqMan™ Fast Advanced Master Mix sufficient for 5000 reactions

TaqMan™ Fast Advanced Master Mix contains AmpliTaq™ Fast DNA Polymerase, Uracil-N glycosylase (UNG), dNTPs (with dUTP), ROX™ dye (passive reference), and optimized buffer.

Store at 2°C to 8°C in the dark.
Detection Method Primer-probe
Form Liquid
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase AmpliTaq Fast DNA Polymerase
Reaction Speed Fast
For Use With (Equipment) 7500 Fast System, 7500 System, 7900HT System, QuantStudio Absolute Q Digital PCR System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 6 Pro, QuantStudio™ 7 Flex, QuantStudio™ 7 Pro, StepOne™, StepOnePlus™, ViiA™ 7 System
Multiplex Capability Duplex
Passive Reference Dye ROX (Pre-mixed)
Product Line TaqMan™
Product Type Fast Advanced Master Mix
Quantity 10 x 5 mL
Shipping Condition Wet Ice
Target Specificity cDNA or gDNA only
For Use With (Application) Gene Expression, DNA Quantitation, Presence/Absence, miRNA analysis
Thermostability 72 hr., Assembled Reaction
Fidelity (vs. Taq) 2 X
No. of Reactions 5000 Reactions
Sample Type DNA (Genomic), cDNA
Volume 50 mL
Includes dUTP, UNG/UDG
Inhibitor Tolerance (Simple) High
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.