Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Argillaceous Shale Specimen
SCI_ED

Catalog No. p-7076841 Shop All Eisco Products
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm

Argillaceous Shale Specimen is formed from cementation and lithification of very small grains of rock, commonly mud or clay.

  • Small clast size indicates deposition in low-energy environment such as shallow lake or slow-moving river
  • For use in sedimentary rock identification exercises due to its diagnostic flat, fissile rock structure
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Small clast size indicates that though the grains had been eroded, rock formed in low energy environment

Specifications

Product Type Sedimentary Rock, Argillaceous Shale
Quantity 10/Pk.
Specimen Size 2 to 3 cm
Weight (Metric) 0.2 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.