Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASB3 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056645T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056645T 0.05 mL
NB10056645 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056645T Supplier Novus Biologicals Supplier No. NB10056645SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ASB3 Polyclonal specifically detects ASB3 in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot.

Specifications

Antigen ASB3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000
Formulation Whole antisera with PBS and 0.1 mg/ml BSA with 0.02% Sodium Azide
Gene Accession No. Q9Y575
Gene Alias ankyrin repeat and SOCS box containing 3, ankyrin repeat and SOCS box protein 3, ankyrin repeat and SOCS box-containing 3, ASB-3MGC132002, FLJ10123, FLJ10421, MGC12531, MGC996
Gene Symbols ASB3
Host Species Rabbit
Immunogen This antibody was developed against synthetic peptides corresponding to amino acids 63-75 and 166-180 of human Asb-3 (NP_665862).
Molecular Weight of Antigen 57.745 kDa
Purification Method Unpurified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 51130
Test Specificity Cross reacts with Human. It does not cross-react with Mouse or Rat Asb-3 .
Target Species Human, Mouse (Negative), Rat (Negative)
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.