Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Ascentis™ Express Peptide ES-C18 HPLC Column, 2.7 μm Particle Size

Catalog No. 111007964 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
3 cm
5 cm
7.5 cm
10 cm
15 cm
Diameter (Metric):
2.1 mm
3 mm
4.6 mm
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
11-100-7964 3 cm 3 mm
11-100-7963 3 cm 2.1 mm
11-100-7543 5 cm 2.1 mm
11-100-7322 7.5 cm 2.1 mm
11-100-7116 10 cm 2.1 mm
11-100-6888 15 cm 2.1 mm
11-100-7544 5 cm 3 mm
11-100-7117 10 cm 3 mm
11-100-6889 15 cm 3 mm
11-100-7965 3 cm 4.6 mm
11-100-7545 5 cm 4.6 mm
11-102-0721 7.5 cm 4.6 mm
11-100-7118 10 cm 4.6 mm
11-101-6072 15 cm 4.6 mm
11-100-7323 7.5 cm 3 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. 11-100-7964 Supplier MilliporeSigma™ Supelco™ Supplier No. 53308U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ascentis™ Express Peptide ES-C18 columns are specifically engineered to separate higher molecular weight compounds such as peptides and small proteins.

Ascentis™ Express Peptide ES-C18 columns contain advanced Fused-Core particles that have larger pores (160 Å versus 90 Å in standard Ascentis™ Express), bonded with sterically-protected C18 ligands to provide extra stability (ES) at very low pH (< 1) and high temperatures (up to 100°C). This greatly expands the application range for Ascentis™ Express columns.
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 3 mm
Length (Metric) 3 cm
Particle Size 2.7 μm
pH 1 to 9
Pore Size 160 Å
Surface Area 80 m2/g
Stationary Phase C18 (octadecyl)
Product Line Ascentis
Matrix C18 (octadecyl) Phase/Fused-Core Particle Platform
Endcapped No
Phase Reversed Phase
Material Stainless Steel
Technique Fused-Core HPLC
Show More Show Less

Ascentis is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.