Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Instrument Check Standard 10 For ICP, SPEX CertiPrep™
SDP

Catalog No. CALMIX10100 Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CALMIX10100 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CALMIX10100 Supplier SPEX CertiPrep Supplier No. CALMIX10100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
K500μg/mL
Al50μg/mL
Ba50μg/mL
Cd50μg/mL
Cu50μg/mL
Mn50μg/mL
Zn50μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 7446-07-3 , 7440-09-7 , 7440-39-3 , 7440-50-8 , 7439-96-5 , 7440-66-6
Molecular Formula O2Te
Molecular Weight (g/mol) 159.60
MDL Number MFCD00011263
InChI Key LAJZODKXOMJMPK-UHFFFAOYSA-N
PubChem CID 62638
IUPAC Name (oxo-λ4-tellanylidene)oxidane
SMILES O=[Te]=O

Specifications

Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Max % 97.92, 0.0, 0.0, 0.0, 0.0, 0.0
Analyte or Component Names Potassium (500mg/L),Aluminum (50mg/L),Barium (50mg/L)
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Element K, Al, Ba
Solvent or Matrix 2% HNO3
Quantity 125 mL
Grade Assurance
Chemical Name or Material Instrument Check Standard 10
Show More Show Less
Hazard Category Danger: Causes severe skin burns and eye damage Do not breathe dusts or mists. IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. If in eyes, rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Hazardous
Recommended Storage Ambient
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.