Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ Astec™ CHIROBIOTIC™ T2 Chiral HPLC Column, 5 μm Particle Size
Shop All MilliporeSigma Supelco Products
Click to view available options
Length (Metric):
10 cm
15 cm
25 cm
Diameter (Metric):
2.1 mm
4.6 mm
Description
CHIROBIOTIC™ T2 columns offer unique selectivity for a number of classes of molecules, specifically underivatized alpha, beta, gamma and cyclic amino acids, N-derivatized amino acids, hydroxy-carboxylic acids, acidic compounds including carboxylic acids and phenols, small peptides, neutral aromatic analytes and cyclic aromatic and aliphatic amines. Separations normally obtained on a chiral crown ether or ligand exchange-type CSPs are also possible on the CHIROBIOTIC™ T and T2, but in much simpler mobile phases, like water-alcohol. In addition, all of the known beta-blockers (amino alcohols), and dihydrocoumarins have been resolved. CHIROBIOTIC™ T and T2 differ in their bonding chemistry and the pore size of the support particle, giving them different selectivity and preparative capacity.
Specifications
Specifications
| Diameter (Metric) | 2.1 mm |
| Length (Metric) | 10 cm |
| Particle Size | 5 μm |
| pH | 3.8 to 6.8 |
| Pore Size | 200 Å |
| Stationary Phase | Teicoplanin Glycopeptide |
| Product Line | Astec CHIROBIOTIC |
| Matrix | Teicoplanin Glycopeptide Phase/High-Purity Silica Gel Particle Platform |
| Phase | Chiral |
| Technique | Chiral HPLC |
Astec and CHIROBIOTIC are registered trademarks of Sigma-Aldrich Co. LLC.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction