Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP6V1G3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | ATP6V1G3 |
---|---|
Dilution | Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:1000-1:2500 |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ATP6V1G3 Polyclonal specifically detects ATP6V1G3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ATP6V1G3 | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
127124 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:AKEEAMVEIDQYRMQRDKEFRLKQSKIMGSQNNLSDEIEEQTLGKIQELNGHYNKYMESVMNQLLSMVCDMKPEIHVNYR | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:1000-1:2500 | |
Polyclonal | |
Rabbit | |
Human | |
ATPase, H+ transporting, lysosomal 13kD, V1 subunit G isoform 3, ATPase, H+ transporting, lysosomal 13kDa, V1 subunit G isoform 3, ATPase, H+ transporting, lysosomal 13kDa, V1 subunit G3, H+ transporting, lysosomal (vacuolar proton pump) subunit G3, MGC119810, MGC119813, vacuolar ATP synthase subunit G 3, vacuolar proton pump G subunit 3, Vacuolar proton pump subunit G 3, vacuolar proton pump, subunit G3, V-ATPase 13 kDa subunit 3, V-ATPase G subunit 3, V-ATPase G3 subunit, V-ATPase subunit G 3, V-type proton ATPase subunit G 3 | |
ATP6V1G3 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title