Learn More
Eisco™ Augite (Pyroxene Mineral) Specimen
SCI_ED
Catalog No. s24673 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S24673 1 Ea.
S26679 10/Pk.
S24674 12/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S24673 Supplier Eisco™ Supplier No. ESNG0011
Only null left
Add to Cart
Add to Cart
Augite (Pyroxene Mineral) Specimen suitable for use in classroom identification lab exercises and for study on 2-plane, 90° cleavage.
  • For lessons on Bowen's reaction series
  • For mineral identification and physical property exercises
  • Handy visual aid during lessons on Bowen's reaction series as well
  • Correlates well to Next Generation Science Standards Evidence Statements
Specifications
Product Type Mineral, Augite
For Use With (Application) Study on Bowen's reaction series
Packaging Clear labeled and barcoded bag
Quantity 1 Ea.
Specimen Size 2 to 3 cm