Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Avian Influenza A H6N1 Nucleoprotein Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 3 publications

$206.50 - $499.50

Specifications

Antigen Avian Influenza A H6N1 Nucleoprotein
Concentration 1.0 mg/ml
Dilution ELISA 0.1-1 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10, Immunohistochemistry-Paraffin 1:10-1:500. Use reported in scientific literature (Thompson et al (2006))
Applications ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056572T
SDP
View Documents
Novus Biologicals
NB10056572SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NB10056572
SDP
View Documents
Novus Biologicals
NB10056572
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

Avian Influenza A H6N1 Nucleoprotein Polyclonal specifically detects Avian Influenza A H6N1 Nucleoprotein in Virus samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications

Specifications

Avian Influenza A H6N1 Nucleoprotein
ELISA 0.1-1 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10, Immunohistochemistry-Paraffin 1:10-1:500. Use reported in scientific literature (Thompson et al (2006))
Polyclonal
Purified
RUO
PBS with 0.05% Sodium Azide
A synthetic peptide corresponding to amino acids 428-441 (AFTGNTEGRTSDMR) of avian flu nucleoprotein were used as the immunogen, GenBank no ADC34563.1.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
1.0 mg/ml
ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Virus, Chicken
NP, Nucleoprotein
IgG
Protein G purified
The amino acid sequence used as immunogen has 100% identity to the H5N1 virus in chicken, duck and swine.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.